missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aconitase 1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-15244
This item is not returnable.
View return policy
Description
Aconitase 1 Polyclonal specifically detects Aconitase 1 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| Aconitase 1 | |
| Polyclonal | |
| Unconjugated | |
| 0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal | |
| Aconitase, aconitase 1, soluble, aconitate hydratase, Citrate hydro-lyase, cytoplasmic aconitate hydratase, EC 4.2.1, EC 4.2.1.3, Ferritin repressor protein, IREB1IREBP1, IREBP, IRE-BP 1, Iron regulatory protein 1, iron-responsive element binding protein 1, Iron-responsive element-binding protein 1, IRP1ACONS | |
| Rabbit | |
| 98 kDa | |
| 0.1 mL | |
| metabolism | |
| 48 | |
| Human | |
| IgG |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| 0.78mg/mL | |
| Western Blot 1:500-1:3000, Immunocytochemistry/Immunofluorescence 1:100-1:1000 | |
| P21399 | |
| ACO1 | |
| Recombinant protein encompassing a sequence within the center region of human Aconitase 1. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human. Expected to cross react based on sequence identity: Rhesus Monkey (100%). | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction