missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ acnB Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA580254
This item is not returnable.
View return policy
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Escherichia coli acnB.
acnB is involved in the catabolism of short chain fatty acids (SCFA) via the tricarboxylic acid (TCA)(acetyl degradation route) and the 2-methylcitrate cycle I (propionate degradation route). It catalyzes the reversible isomerization of citrate to isocitrate via cis-aconitate. Also catalyzes the hydration of 2-methyl-cis-aconitate to yield (2R,3S)-2-methylisocitrate. The apo form of AcnB functions as a RNA-binding regulatory protein. During oxidative stress inactive AcnB apo-enzyme without iron sulfur clusters binds the acnB mRNA 3' UTRs (untranslated regions), stabilizes acnB mRNA and increases AcnB synthesis, thus mediating a post-transcriptional positive autoregulatory switch. AcnB also decreases the stability of the sodA transcript.
Specifications
| acnB | |
| Polyclonal | |
| Unconjugated | |
| ACN; Aconitase | |
| Recombinant Escherichia coli acnB protein. | |
| 100 μL | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA | |
| 1 mg/mL | |
| PBS with no preservative | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| Bacteria | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction