missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ACMSD Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5113272
This item is not returnable.
View return policy
Description
ACMSD Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA
The neuronal excitotoxin quinolinate is an intermediate in the de novo synthesis pathway of NAD from tryptophan, and has been implicated in the pathogenesis of several neurodegenerative disorders. Quinolinate is derived from alpha-amino-beta-carboxy-muconate-epsilon-semialdehyde (ACMS). ACMSD (ACMS decarboxylase; EC 4.1.1.45) can divert ACMS to a benign catabolite and thus prevent the accumulation of quinolinate from ACMS.[supplied by OMIM, Oct 2004].
Specifications
| ACMSD | |
| Polyclonal | |
| Unconjugated | |
| ACMSD | |
| 2-amino-3-carboxylmuconate-6-semialdehyde decarboxylase; 2-amino-3-carboxymuconate-6-semialdehyde decarboxylase; Acmsd; amino carboxymuconate semialdehyde decarboxylase; aminocarboxymuconate semialdehyde decarboxylase; Picolinate carboxylase; zgc:162888 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 130013, 171385 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 4 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q8R5M5, Q8TDX5 | |
| ACMSD | |
| Recombinant Human 2-amino-3-carboxymuconate-6-semialdehyde decarboxylase protein (33-129AA). | |
| 100 μg | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction