missing translation for 'onlineSavingsMsg'
Learn More

Acitretin, MedChemExpress

Product Code. 16441376
Encompass_Preferred
Click to view available options
Quantity:
100 mg
200 mg
500 mg
Unit Size:
100mg
200mg
500mg
This item is not returnable. View return policy

Product Code. 16441376

Brand: Medchem Express HYB0107

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Acitretin (Ro 10-1670) is a second-generation, systemic retinoid that has been used in the treatment of psoriasis. Acitretin also can be used for the research of Alzheimer’s disease.

Acitretin (Ro 10-1670) is a second-generation, systemic retinoid that has been used in the treatment of psoriasis. Acitretin also can be used for the research of Alzheimer’s disease.

Specifications

Chemical Name or Material Acitretin
CAS 55079-83-9
Color Yellow
Purity Grade Notes Research
Molecular Formula C21H26O3
Quantity 200 mg
Synonym Ro 10-1670
Solubility Information DMSO : 20 mg/mL (61.27 mM; Need ultrasonic) ∣H2O : 0.67 mg/mL (2.05 mM; Need ultrasonic)
SMILES CC(/C=C/C(C(C)=CC(OC)=C1C)=C1C)=C\C=C\C(C)=C\C(O)=O
Molecular Weight (g/mol) 326.43
Formula Weight 326.43
Percent Purity 99.66%
Grade Research
Physical Form Powder
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.