missing translation for 'onlineSavingsMsg'
Learn More

Acipimox, MedChemExpress

Product Code. 16428216
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16428216 100 mg 100mg
16414327 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16428216 Supplier Medchem Express Supplier No. HYB0283
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Acipimox (K-9321), a nicotinic acid analogue, is an antilipolytic compound. Acipimox stimulates leptin releas, inhibits lipolysis and suppresses systemic levels of free fatty acids (FFAs) and improves insulin sensitivity.

Acipimox (K-9321), a nicotinic acid analogue, is an antilipolytic compound. Acipimox stimulates leptin releas, inhibits lipolysis and suppresses systemic levels of free fatty acids (FFAs) and improves insulin sensitivity.

Specifications

Chemical Name or Material Acipimox
CAS 51037-30-0
Color Off-White
Purity Grade Notes Research
Molecular Formula C6H6N2O3
Quantity 100 mg
Synonym K-9321
Solubility Information DMSO : ≥ 100 mg/mL (648.85 mM) ∣H2O : 20 mg/mL (129.77 mM; Need ultrasonic)
SMILES O=C(C1=C[N+]([O-])=C(C)C=N1)O
Molecular Weight (g/mol) 154.12
Formula Weight 154.12
Percent Purity 99.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.