missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Acinus Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA517344
This item is not returnable.
View return policy
Description
It is not recommended to aliquot this antibody.
ACINUS (apoptotic chromatin condensation inducer in the nucleus) is an induced of chromatin condensation. ACINUS is cleaved by caspase-3 and an additional unknown protease generating a small active peptide, p17, which causes chromatin condensation in vitro when it is added to purified nuclei. ACINUS also induces apoptotic chromatin condensation in cells. ACINUS is ubiquitously expressed. Three different spliced forms of Acinus have been identified in human and mouse.
Specifications
| Acinus | |
| Polyclonal | |
| Unconjugated | |
| ACIN1 | |
| 2610036I19Rik; 2610510L13Rik; Acin1; ACINUS; acinusL; acinusS; Acn; apoptotic chromatin condensation inducer 1; apoptotic chromatin condensation inducer in the nucleus; C79325; DKFZp667N107; fSAP152; functional spliceosome-associated protein 152; KIAA0670; mKIAA0670 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 22985, 305884, 56215 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 35.1 μg/mL | |
| 0.01M HEPES with 100μg/mL BSA, 50% glycerol, 0.15M NaCl and no preservative; pH 7.5 | |
| Q9JIX8, Q9UKV3 | |
| ACIN1 | |
| Synthetic peptide corresponding to residues surrounding Ala835 of Acinus. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat, Monkey | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction