missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Acinus Polyclonal Antibody

Product Code. 12828162
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12828162 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12828162 Supplier Invitrogen™ Supplier No. PA519948

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

In Western blot applications, this antibody detects a band at ~220kDa. A suggested positive control is K562 cell lysate. PA5-19948 can be used with blocking peptide PEP-0073.

ACINUS (apoptotic chromatin condensation inducer in the nucleus) is an induced of chromatin condensation. ACINUS is cleaved by caspase-3 and an additional unknown protease generating a small active peptide, p17, which causes chromatin condensation in vitro when it is added to purified nuclei. ACINUS also induces apoptotic chromatin condensation in cells. ACINUS is ubiquitously expressed. Three different spliced forms of Acinus have been identified in human and mouse.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Acinus
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene ACIN1
Gene Accession No. Q9UKV3
Gene Alias 2610036I19Rik; 2610510L13Rik; Acin1; ACINUS; acinusL; acinusS; Acn; apoptotic chromatin condensation inducer 1; apoptotic chromatin condensation inducer in the nucleus; C79325; DKFZp667N107; fSAP152; functional spliceosome-associated protein 152; KIAA0670; mKIAA0670
Gene Symbols ACIN1
Host Species Rabbit
Immunogen A peptide corresponding to amino acids 994 to 1009 of human AcinusL, 267 to 282 of human AcinusS prime, or 236 to 251 of human AcinusS.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22985
Target Species Human
Content And Storage Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.