missing translation for 'onlineSavingsMsg'
Learn More

Acifluorfen-methyl-2-amino, TRC

Product Code. 30006857
Change view
Click to view available options
Quantity:
50 mg
500 mg
Unit Size:
500mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30006857 50 mg 50mg
30011987 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30006857 Supplier Toronto Research Chemicals Supplier No. TRCA18997550MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58105-66-1
Molecular Formula C15 H11 Cl F3 N O3
Molecular Weight (g/mol) 345.7
Synonym Benzoic acid, 2-amino-5-[2-chloro-4-(trifluoromethyl)phenoxy]-, methyl ester, 4-Amino-3-carbomethoxy-2'-chloro-4'-trifluoromethyldiphenyl ether
IUPAC Name methyl 2-amino-5-[2-chloro-4-(trifluoromethyl)phenoxy]benzoate
SMILES COC(=O)c1cc(Oc2ccc(cc2Cl)C(F)(F)F)ccc1N

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C15H11ClF3NO3/c1-22-14(21)10-7-9(3-4-12(10)20)23-13-5-2-8(6-11(13)16)15(17,18)19/h2-7H,20H2,1H3
Formula Weight 345.038
Chemical Name or Material Acifluorfen-methyl-2-amino
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.