missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acidic Calponin Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£381.00
Specifications
| Antigen | Acidic Calponin |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
Acidic Calponin Polyclonal specifically detects Acidic Calponin in Human samples. It is validated for Western Blot.Specifications
| Acidic Calponin | |
| Polyclonal | |
| Rabbit | |
| Q15417 | |
| 1266 | |
| Synthetic peptides corresponding to CNN3 (calponin 3, acidic) The peptide sequence was selected from the C terminal of CNN3. Peptide sequence GLGRQVYDPKYCAAPTEPVIHNGSQGTGTNGSEISDSDYQAEYPDEYHGE. | |
| Primary |
| Western Blot | |
| Unconjugated | |
| RUO | |
| calponin 3, acidic, Calponin, acidic isoform, calponin-3, dJ639P13.2.2 (acidic calponin 3) | |
| CNN3 | |
| IgG | |
| 36 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title