missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acid Surrogate Standard, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
£62.50 - £97.20
Specifications
Chemical Name or Material | Acid Surrogate Standard |
---|---|
CAS | 118-79-6, 93951-73-6, 367-12-4, 13127-88-3 |
Concentration or Composition Notes | Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity. |
For Use With (Equipment) | GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation |
Concentration or Composition (by Analyte or Components) | 2000μg/mL |
Includes: Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
12749695
|
SPEX Certiprep
CLP90-SA |
1 mL | 2mL amber prescored glass ampule | ||||||
12759695
|
SPEX Certiprep
CLP90-SA5 |
5 mL | 5mL amber prescored glass ampule | ||||||
15276366
|
SPEX Certiprep
CLP90-SA5 |
5 mL | 5mL amber prescored glass ampule |
£64.85
5mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
Analyte | CAS No. | Concentration |
---|---|---|
2,4,6-Tribromophenol | 118-79-6 | 2000μg/mL |
2-Chlorophenol-3,4,5,6-d4 | 93951-73-6 | 2000μg/mL |
2-Fluorophenol | 367-12-4 | 2000μg/mL |
Phenol-d6 | 13127-88-3 | 2000μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
Acid Surrogate Standard | |
Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity. | |
2000μg/mL | |
Methanol |
118-79-6, 93951-73-6, 367-12-4, 13127-88-3 | |
GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation | |
2,4,6-Tribromophenol |
Safety and Handling
Harmful if swallowed. Suspected of causing cancer.
missing translation for 'dotInformation' : Hazardous
Recommended Storage : Ambient
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Danger
Hazard Category
- Acute toxicity Category 3
- Acute toxicity Category 4
- Flammable liquid Category 2
- Specific target organ toxicity after single exposure Category 1
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H302-Harmful if swallowed.
- H331-Toxic if inhaled.
- H370-Causes damage to organs.
Precautionary Statement
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P103-Read label before use.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : methanol
Product intended for laboratory use only