missing translation for 'onlineSavingsMsg'
Learn More

Acid Red 26 (>80%), TRC

Product Code. 30323626
Click to view available options
Quantity:
1 g
Unit Size:
1g
This item is not returnable. View return policy

Product Code. 30323626

Brand: Toronto Research Chemicals TRCA1898451G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Acid Red 26
CAS 3761-53-3
Molecular Formula C18 H14 N2 O7 S2 . 2 Na
Quantity 1 g
Synonym 2,7-Naphthalenedisulfonic acid, 4-[(2,4-dimethylphenyl)azo]-3-hydroxy-, disodium salt (9CI), C.I. Acid Red 26 (7CI), C.I. Acid Red 26, disodium salt (8CI), Ponceau Xylidine (6CI), 1-(2,4-Xylylazo)-2-naphthol-3,6-disulfonic acid disodium salt, 1695 Red, Acid Leather Red KPR, Acid Leather Red P 2R, Acid Leather Scarlet IRW, Acid Ponceau 2RL, Acid Ponceau R, Acid Ponceau Special, Acid Red 26, Acid Scarlet, Acid Scarlet 2B, Acid Scarlet 2R, Acid Scarlet 2RL, Acidal Ponceau G, Ahcocid Fast Scarlet R, Aizen Ponceaux RH, Amacid Lake Scarlet 2R, Borunil Ponceau A 2R, Brilliant Ponceau G, C.I. 16150, C.I. Food Red 5, Calcocid Scarlet 2R, Calcolake Scarlet 2R, Certicol Ponceau MXS, Colacid Ponceau Special, Colorosacid Scarlet, D and C Red No. 5, Disodium salt of 1-(2,4-xylylazo)-2-naphthol-3,6-disulfonic acid, Edicol Supra Ponceau R, Fenazo Scarlet 2R, Food Red 5, Hexacol Ponceau 2R, Hexacol Ponceau MX, Hidacid Scarlet 2R, Japan Red 503, Japan Red No. 503, Kiton Ponceau 2R, Kiton Ponceau R, Kiton Scarlet 2RC, Lake Scarlet 2RBN, Lake Scarlet R, Naphthalene Lake Scarlet R, Naphthalene Scarlet R, Naphthazine Scarlet 2R, Naphthazine Scarlet 2RN, Neklacid Red RR, New Ponceau 4R, Paper Red HRR, Pigment Ponceau R, Ponceau 2R, Ponceau 2R Extra A Export, Ponceau 2RL, Ponceau 2RX, Ponceau BNA, Ponceau G, Ponceau MX, Ponceau PXM, Ponceau R, Ponceau RR, Ponceau RR Type 8019, Ponceau RS, Ponceau Red R, Ponsol R, Red No.503, Scarlet 2R, Scarlet 2RB, Scarlet 2RL Bluish, Scarlet R, Scarlet RRA, Tertracid Ponceau 2R, Triacid Ponceau 2R, Xylidine Ponceau, Xylidine Ponceau 2R, Xylidine Red
InChI Formula InChI=1S/C18H16N2O7S2.2Na/c1-10-3-6-15(11(2)7-10)19-20-17-14-5-4-13(28(22,23)24)8-12(14)9-16(18(17)21)29(25,26)27;;/h3-9,21H,1-2H3,(H,22,23,24)(H,25,26,27);;/q;2*+1/p-2
SMILES [Na+].[Na+].Cc1ccc(N=Nc2c(O)c(cc3cc(ccc23)S(=O)(=O)[O-])S(=O)(=O)[O-])c(C)c1
IUPAC Name disodium;4-[(E)-(2,4-dimethylphenyl)diazenyl]-3-hydroxynaphthalene-2,7-disulfonate
Molecular Weight (g/mol) 480.42
Formula Weight 480.0038
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.