missing translation for 'onlineSavingsMsg'
Learn More

Acid Red 114 (Technical Grade), TRC

Product Code. 30132987
Change view
Click to view available options
Quantity:
500 mg
1 g
2.5 g
Unit Size:
1g
2.5g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30132987 2.5 g 2.5g
30040840 500 mg 500mg
17907829 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30132987 Supplier Toronto Research Chemicals Supplier No. TRCA7952632.5G
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6459-94-5
Molecular Formula C37 H28 N4 O10 S3 . 2 Na
Molecular Weight (g/mol) 830.81
Synonym 1,3-Naphthalenedisulfonic acid, 8-[[3,3'-dimethyl-4'-[[4-[[(4-methylphenyl)sulfonyl]oxy]phenyl]azo][1,1'-biphenyl]-4-yl]azo]-7-hydroxy-, disodium salt (9CI), C.I. Acid Red 114 (7CI), C.I. Acid Red 114, disodium salt (8CI), Acid Leather Red BG, Acid Milling Red BS, Acid Milling Red RS, Acid Red 114, Acid Red F-RS, Acid Red P-RS, Acid Red RS, Amacid Milling Red PRS, Anadurm Red M-R, Apollo Nylon Fast Red R, Atul Acid Milling Red RS, Benzyl Fast Red BG, Benzyl Red BR, Best Acid Milling Red RS, C.I. 23635, Colomill Red RS, Concorde Acid Red M-RS, Concorde Leather Red RSN, Covalene Red RS, Covalene Scarlet RS, Covanyl Scarlet RS, Daedo Acid Red RS, Dinacid Milling Red RG, Dycosweak Acid RS, Eniacid Fast Red R, Erionyl Red RS, Erionyl Red RS 125, Everacid Milling Red RS, Everlan Red RS, Fabracid Red M-RS, Fenafor Red PB, Folan Red B, Indacid Milling Red RS, Kayanol Milling Red RS, Kayanol Milling Red RS 125, Kenamide Red K 2R, Kenanthrol Red R, Leather Fast Red B, Lerui Acid Red F-RS, Levanol Red GG, Midlon Red PRS, Milling Fast Red B, Milling Fast Red R, Milling Red B, Milling Red BB, Milling Red SWB, Monacid Red RS, Multicuer Red R, Navimill Red RS, Navitan Red RS, Pacid Red RS, Polar Red RS, Ratna Acid Red RS, Rifa Acid Fast Red RS, Rifa Leather Red R, Sandolan Red N-RS, Sandolan Red N-RSI, Sandolan Red RSNI, Sella Fast Red RS, Simacid Red 24900, Sulphonol Red R, Suminol Milling Red RS, Suminol Milling Red RS 125, Supranol Fast Red 3G, Supranol Fast Red GG, Supranol Red PBX-CF, Supranol Red R, Telon Fast Red GG, Tertracid Milling Red B, Triacid Fast Red RS, Vondamol Fast Red RS
IUPAC Name disodium;7-hydroxy-8-[(E)-[2-methyl-4-[3-methyl-4-[(E)-[4-(4-methylphenyl)sulfonyloxyphenyl]diazenyl]phenyl]phenyl]diazenyl]naphthalene-1,3-disulfonate
SMILES [Na+].[Na+].Cc1ccc(cc1)S(=O)(=O)Oc2ccc(cc2)N=Nc3ccc(cc3C)c4ccc(N=Nc5c(O)ccc6cc(cc(c56)S(=O)(=O)[O-])S(=O)(=O)[O-])c(C)c4

Specifications

Quantity 2.5 g
InChI Formula InChI=1S/C37H30N4O10S3.2Na/c1-22-4-13-30(14-5-22)54(49,50)51-29-11-9-28(10-12-29)38-39-32-15-6-25(18-23(32)2)26-7-16-33(24(3)19-26)40-41-37-34(42)17-8-27-20-31(52(43,44)45)21-35(36(27)37)53(46,47)48;;/h4-21,42H,1-3H3,(H,43,44,45)(H,46,47,48);;/q;2*+1/p-2
Formula Weight 830.0763
Chemical Name or Material Acid Red 114
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.