missing translation for 'onlineSavingsMsg'
Learn More

Acid Red 1, Thermo Scientific™

Product Code. 17790619 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17790619 5 g 5g
17700629 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17790619 Supplier Thermo Scientific Acros Supplier No. 198700050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Absorbance ≥250 lambda 503 - 509 nm, 0.02 g/l water; >250 lambda 530 - 534 nm, 0.02 g/l water
Chemical Name or Material Acid Red 1
CAS 3734-67-6
Molecular Formula C18 H13 N3 Na2 O8 S2
MDL Number 00003954
Synonym 5-Acetamido-4-hydroxy-3-phenylazo-2,7-naphthalene-disulfonicacid, disodium salt; Amido Naphthol Red G; Azophloxine; C.I. 18050
Solubility Information Soluble in water
Infrared Spectrum Conforms
Molecular Weight (g/mol) 509.43
Loss on Drying ≤10 %
Packaging Glass bottle
Certificate of Analysis Available
Color Purple to Red
Quantity 5 g
Physical Form Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.