missing translation for 'onlineSavingsMsg'
Learn More

EKASTU Safety Acid Proof Hood for C 607 und SFERA

Product Code. 10476981
Change view
Click to view available options
Color:
Yellow
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
10476981 Yellow Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10476981 Supplier EKASTU Safety Supplier No. 666971
 Restricted Item
Estimated Shipment: 01-04-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

EKASTU Acid Proof Hood for C 607/SFERA covers the head and neck and features a mask aperture with lead-proof collar

Specifications

Product Type Acid-proof Hood
Size Universal
Material PVC-coated Polyester
Color Yellow
Disposable Reusable
For Use With (Equipment) Full Face Mask C 607/Selecta (Class 2), Full Face Mask SFERA (Class 3)
Quantity 1 Ea.
Dimensions (L x W x H) 25.5 x 3.5 x 39 cm
Weight (Metric) Approx. 296 g
Hood Type Acid-proof hood
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.