missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Acid Phosphatase 1 Recombinant Rabbit Monoclonal Antibody (PSH04-57)

Rabbit Recombinant Monoclonal Antibody
Marke: Invitrogen™ MA556401
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Acid Phosphatase 1 Recombinant Monoclonal Antibody for Western Blot, IHC (P)
Low molecular weight protein tyrosine phosphtases, including the human red cell phosphatase, HCPTP, are widely expressed cytosolic proteins of approximately 18 kDa that exist in distinct isoforms and are highly selective for phosphotyrosine over phosphoserine or phosphotyrosine. The HCPTP-A and HCPTP-B proteins expressed in human red cells and placenta are fast and slow forms of red cell acid phosphatase5. Like other tyrosine phosphatases, HCPTP is sensitive to inhibition by vanadate, and has a catalytic mechanism that involves formation of a cystinylphosphate intermediate6. The crystal structure of the bovine heart derived enzyme BHPTP shows a four-stranded central parallel b sheet with flanking a helices and an active site cysteine residue in the typical tyrosine phosphatase sequence context, CXXXXXR.
Spezifikation
| Acid Phosphatase 1 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with 0.1% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| P24666 | |
| ACP1 | |
| Recombinant protein within human ACP1 aa 1-158/158. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Western Blot | |
| PSH04-57 | |
| Unconjugated | |
| ACP1 | |
| 4632432E04Rik; acid phosphatase; acid phosphatase 1; acid phosphatase 1, soluble; acid phosphatase of erythrocyte; acp1; Acp-1; Acp1-PA; adipocyte acid phosphatase; Adult cuticle protein 1; AI427468; CG7216; CG7216-PA; cytoplasmic phosphotyrosyl protein phosphatase; Dacp-1; Dmel\CG7216; Dmel_CG7216; HAAP; HAAP betaÊ; hypothetical protein; LMW-PTP; LMW-PTPase; LMW-PTP-I low molecular weight phosphotyrosine protein phosphatase isozyme 1; Low molecular weight cytosolic acid phosphatase; low molecular weight phosphotyrosine protein phosphatase; MGC111030; MGC3499; phosphatase 1, soluble; protein tyrosine phosphatase; ptpase; RCJMB04_11a4; Red cell acid phosphatase 1; RH03221p; testicular secretory protein Li 37; Unknown (protein for MGC:128334) | |
| Rabbit | |
| Protein A | |
| RUO | |
| 52 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur