missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acid Phosphatase 1 Polyclonal Antibody

Acid Phosphatase 1 Polyclonal Antibody
Brand: Invitrogen PA526418
This item is not returnable.
View return policy
Description
Acid Phosphatase 1 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow
Specifications
| Acid Phosphatase 1 | |
| Polyclonal | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| P24666 | |
| ACP1 | |
| KLH conjugated synthetic peptide between 33-61 amino acids from the N-terminal region of human ACP1. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| Unconjugated | |
| ACP1 | |
| 4632432E04Rik; acid phosphatase; acid phosphatase 1; acid phosphatase 1, soluble; acid phosphatase of erythrocyte; acp1; Acp-1; Acp1-PA; adipocyte acid phosphatase; Adult cuticle protein 1; AI427468; CG7216; CG7216-PA; cytoplasmic phosphotyrosyl protein phosphatase; Dacp-1; Dmel\CG7216; Dmel_CG7216; HAAP; HAAP betaÊ; hypothetical protein; LMW-PTP; LMW-PTPase; LMW-PTP-I low molecular weight phosphotyrosine protein phosphatase isozyme 1; Low molecular weight cytosolic acid phosphatase; low molecular weight phosphotyrosine protein phosphatase; MGC111030; MGC3499; phosphatase 1, soluble; protein tyrosine phosphatase; ptpase; RCJMB04_11a4; Red cell acid phosphatase 1; RH03221p; testicular secretory protein Li 37; Unknown (protein for MGC:128334) | |
| Rabbit | |
| Antigen Affinity Chromatography, Protein A | |
| RUO | |
| 52 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction