missing translation for 'onlineSavingsMsg'
Learn More

Acid Number Standard 0.1 mgKOH/g. Reference materials intended for use in the determination of Acid, VHG™

Product Code. 17901116
Change view
Click to view available options
Quantity:
100 g
400 g
800 g
Unit Size:
100g
400g
800g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17901116 100 g 100g
17992236 800 g 800g
17963926 400 g 400g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17901116 Supplier VHG Labs Supplier No. VHGAN0.1100G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

Acid Number Standard 0.1 mgKOH/g. Reference materials intended for use in the determination of Acid
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Potassium hydroxide
CAS 1310-58-3
Methods Potentiometric Titration
Analyte or Component Names Potassium hydroxide
Shipping Condition Room Temperature
Molecular Formula H K O
Certifications/Compliance ISO 17034
Synonym Potassium hydroxide
InChI Formula InChI=1S/K.H2O/h;1H2/q+1;/p-1
SMILES O[K]
IUPAC Name hydroxypotassium
Solvent or Matrix Hydrocarbon Oil
Molecular Weight (g/mol) 56.11
Quantity 100 g
Formula Weight 55.9664 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.