missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acid Number 0.1 mgKOH/g Reference Standard in Hydrocarbon Oil
Specifications
Specifications
| Chemical Name or Material | Acid Number |
| Analyte or Component Names | Acid Number |
| Shipping Condition | Room Temperature |
| Concentration | 0.1 mgKOH/g |
| Certifications/Compliance | ISO 17034 |
| Solution Type | Single Solution |
| Solvent or Matrix | Heavy Mineral Oil |
| Industry Type | Industrial QC |
| Quantity | 50 g |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?