missing translation for 'onlineSavingsMsg'
Learn More

Acid fuchsin sodium salt, MP Biomedicals™

Product Code. 11405611 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25 g
50 g
100 g
Unit Size:
100g
25g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11405611 25 g 25g
11415611 50 g 50g
11425611 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11405611 Supplier MP Biomedicals Supplier No. 0215819725
 Restricted Item
Estimated Shipment: 07-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Acid Fuchsin is one of the dyes used in Masson's Trichome Stain where it is used to distinguish muscle from collagen

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3244-88-0
Molecular Formula C20H19N3Na2O9S3
Molecular Weight (g/mol) 587.544
InChI Key VCEFFPMSOGDMJX-UHFFFAOYSA-N
Synonym acid fuchsin, acidal fuchsine, acid fuchsine, acidal magenta, fuchsine acid, fuchsin acid, p-fuchsine acid, acid fuchsine n, acid fuchsine o, acid fuchsine s
PubChem CID 131852436
IUPAC Name 3-[bis(4-amino-3-sulfophenyl)methylidene]-6-imino-5-methylcyclohexa-1,4-diene-1-sulfonic acid;sodium
SMILES CC1=CC(=C(C2=CC(=C(C=C2)N)S(=O)(=O)O)C3=CC(=C(C=C3)N)S(=O)(=O)O)C=C(C1=N)S(=O)(=O)O.[Na].[Na]

Specifications

Color Black-Blue to Black-Green
Quantity 25 g
Formula Weight 585.5
Vapor Pressure 0
Physical Form Powder
Chemical Name or Material Acid fuchsin sodium salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.