missing translation for 'onlineSavingsMsg'
Learn More

Acid Blue 3 Sodium Salt (>90%), TRC

Product Code. 30325323
Click to view available options
Quantity:
10 g
Unit Size:
10g
This item is not returnable. View return policy

Product Code. 30325323

Brand: Toronto Research Chemicals TRCA18981010G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Patent Blue 5 (E131)
CAS 20262-76-4
Molecular Formula C27 H31 N2 O7 S2 . Na
Quantity 10 g
Synonym Ethanaminium, N-[4-[[4-(diethylamino)phenyl](5-hydroxy-2,4-disulfophenyl)methylene]-2,5-cyclohexadien-1-ylidene]-N-ethyl-, inner salt, sodium salt (1:1), C.I. Acid Blue 3, monosodium salt (8CI), Ethanaminium, N-[4-[[4-(diethylamino)phenyl](5-hydroxy-2,4-disulfophenyl)methylene]-2,5-cyclohexadien-1-ylidene]-N-ethyl-, hydroxide, inner salt, monosodium salt, Ethanaminium, N-[4-[[4-(diethylamino)phenyl](5-hydroxy-2,4-disulfophenyl)methylene]-2,5-cyclohexadien-1-ylidene]-N-ethyl-, inner salt, monosodium salt (9CI), C.I. 42051:2, C.I. Food Blue 5:2, Food Blue 5:2, Patent Blue, Patent Blue V, Patent Blue V Sodium Salt, N-[4-[[4-(Diethylamino)phenyl](5-hydroxy-2,4-disulfophenyl)methylene]-2,5-cyclohexadien-1-ylidene]-N-ethylethanaminium monosodium salt, Acid Blue 3 monosodium salt
InChI Formula InChI=1S/C27H32N2O7S2.Na/c1-5-28(6-2)21-13-9-19(10-14-21)27(20-11-15-22(16-12-20)29(7-3)8-4)23-17-24(30)26(38(34,35)36)18-25(23)37(31,32)33;/h9-18H,5-8H2,1-4H3,(H2-,30,31,32,33,34,35,36);/q;+1/p-1
SMILES [Na+].CCN(CC)c1ccc(cc1)C(=C2C=CC(=[N+](CC)CC)C=C2)c3cc(O)c(cc3S(=O)(=O)[O-])S(=O)(=O)[O-]
IUPAC Name sodium;4-[[4-(diethylamino)phenyl]-(4-diethylazaniumylidenecyclohexa-2,5-dien-1-ylidene)methyl]-6-hydroxybenzene-1,3-disulfonate
Molecular Weight (g/mol) 582.66
Formula Weight 582.147
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.