missing translation for 'onlineSavingsMsg'
Learn More

Acid Blue 29, MP Biomedicals™

Product Code. 11477922 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11477922 25 g 25g
11487922 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11477922 Supplier MP Biomedicals Supplier No. 0215259625
 Restricted Item
Estimated Shipment: 02-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Stains and dyes

Chemical Identifiers

CAS 5850-35-1
Molecular Formula C22H14N6Na2O9S2
Molecular Weight (g/mol) 616.49
MDL Number MFCD00009960
InChI Key WEILTBATXPPXPP-AXSRUCDFSA-L
Synonym C.I. 20460
PubChem CID 44134491
IUPAC Name disodium (3E)-5-amino-6-[2-(3-nitrophenyl)diazen-1-yl]-4-oxo-3-(2-phenylhydrazin-1-ylidene)-3,4-dihydronaphthalene-2,7-disulfonate
SMILES [Na+].[Na+].NC1=C(N=NC2=CC=CC(=C2)[N+]([O-])=O)C(=CC2=C1C(=O)\C(=N/NC1=CC=CC=C1)C(=C2)S([O-])(=O)=O)S([O-])(=O)=O

Specifications

Formula Weight 616.5
Color Black
Quantity 25 g
Physical Form Powder
Chemical Name or Material Acid Blue 29
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.