missing translation for 'onlineSavingsMsg'
Learn More

ACHP Hydrochloride, MedChemExpress

Product Code. 16438237
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16438237

Brand: Medchem Express HY13060

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress ACHP Hydrochloride (IKK-2 Inhibitor VIII) is a highly potent and selective IKK-β inhibitor with an IC50 of 8.5 nM.

ACHP Hydrochloride (IKK-2 Inhibitor VIII) is a highly potent and selective IKK-β inhibitor with an IC50 of 8.5 nM.

Specifications

Chemical Name or Material ACHP Hydrochloride
CAS 406209-26-5
Color Yellow
Purity Grade Notes Research
Molecular Formula C21H25ClN4O2
Quantity 5 mg
Synonym IKK-2 Inhibitor VIII
Solubility Information DMSO : 50 mg/mL (124.72 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble)
SMILES N#CC1=C(C=C(N=C1N)C2=C(C=CC=C2OCC3CC3)O)C4CCNCC4.[H]Cl
Molecular Weight (g/mol) 400.9
Formula Weight 400.9
Percent Purity 98.1%
Grade Research
Physical Form Powder
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.