missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACF1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
£222.00 - £470.00
Specifications
| Antigen | ACF1 |
|---|---|
| Dilution | Immunohistochemistry 1:20 - 1:50, Immunohistochemistry-Paraffin 1:20 - 1:50 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18436001
|
Novus Biologicals
NBP1-90270-25ul |
25 μL |
£222.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18228367
|
Novus Biologicals
NBP1-90270 |
0.1 mL |
£470.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ACF1 Polyclonal specifically detects ACF1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| ACF1 | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 11177 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:RGHRESALKETLLQEKSRICAQLARFSEEKFHFSDKPQPDSKPTYSRGRSSNAYDPSQMCAEKQLELRLRDFLLDIEDRIYQGTLGAIKVTDRHIWRSALESGRYELLSEENKENGIIKTVNEDVEEMEIDEQTK | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry 1:20 - 1:50, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| ACF1hWALp1, ATP-dependent chromatin remodeling protein, ATP-dependent chromatin-remodeling protein, ATP-utilizing chromatin assembly and remodeling factor 1, bromodomain adjacent to zinc finger domain protein 1A, bromodomain adjacent to zinc finger domain, 1A, CHRAC subunit ACF1, hACF1FLJ14383, WALp1, WCRF180DKFZp586E0518, Williams syndrome transcription factor-related chromatin-remodeling factor 180 | |
| BAZ1A | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title