missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ ACF 293T Cell Transient Overexpression Lysate

Product Code. 18393329 Shop All Bio Techne Products
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.01mL
This item is not returnable. View return policy

Product Code. 18393329

Brand: Novus Biologicals™ H00029974T01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Overexpression Lysate

TRUSTED_SUSTAINABILITY

Specifications

Quantity 0.1 mL
Content And Storage Store at -80°C. Avoid freeze-thaw cycles.
Target ACF
Synonym ACF65, ACFASPACF64, apo-B RNA editing protein, APOBEC1 complementation factor, apobec-1 complementation factor (ACF) (ASP), APOBEC-1 stimulating protein, APOBEC1CF, APOBEC1-stimulating protein, MGC163391
Gene Symbol A1CF
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.