missing translation for 'onlineSavingsMsg'
Learn More

Acetylsalicylsalicylic Acid, TRC

Product Code. 17932685
Change view
Click to view available options
Quantity:
100 mg
250 mg
500 mg
Unit Size:
100mg
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17932685 100 mg 100mg
17982875 500 mg 500mg
17992125 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17932685 Supplier Toronto Research Chemicals Supplier No. TRCA187788100MG
 Restricted Item
Estimated Shipment: 10-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 530-75-6
Molecular Formula C16 H12 O6
Molecular Weight (g/mol) 300.26
Synonym 2-[[2-(Acetyloxy)benzoyl]oxy]benzoic Acid, Carbasalate Calcium Imp. B (EP), Acetylsalicylic Acid Imp. D (EP), Acetylsalicylsalicylic Acid
IUPAC Name 2-(2-acetyloxybenzoyl)oxybenzoic acid
SMILES CC(=O)Oc1ccccc1C(=O)Oc2ccccc2C(=O)O

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C16H12O6/c1-10(17)21-14-9-5-3-7-12(14)16(20)22-13-8-4-2-6-11(13)15(18)19/h2-9H,1H3,(H,18,19)
Formula Weight 300.0634
Percent Purity >95%
Chemical Name or Material Acetylsalicylsalicylic acid
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.