missing translation for 'onlineSavingsMsg'
Learn More

Acetylsalicylic Acid, TRC

Product Code. 30328080
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30328080 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30328080 Supplier Toronto Research Chemicals Supplier No. TRCA187780250MG
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Acetylsalicylic Acid
CAS 50-78-2
Purity Grade Notes HPLC
Molecular Formula C9 H8 O4
Quantity 250 mg
Synonym Rhodine (7CI), Salicylic acid acetate (8CI), 2-(Acetyloxy)benzoic acid, 2-Acetoxybenzoic acid, 2-Carboxyphenyl acetate, A.S.A. Empirin, AC 5230, ASA, Acenterine, Acesal, Acesan, Acetard, Aceticyl, Acetilum acidulatum, Acetisal, Acetol, Acetonyl, Acetophen, Acetosal, Acetosalic acid, Acetosalin, Acetylin, Acetylsal, Acetyonyl, Acetysal, Acidum acetylsalicylicum, Acimetten, Acisal, Acylpyrin, Adiro, Albyl E, Angettes 75, Anopyrin, Asaflow, Asagran, Asatard, Ascoden 30, Ascolong, Ascriptin, Aspalon, Aspergum, Aspirdrops, Aspirin, Aspirin Protect 100, Aspirin Protect 300, Aspirin-Direkt, Aspirina, Aspirina 03, Aspirine, Aspro, Aspro Clear, Aspropharm, Asteric, Astrix, Bayaspirin, Benaspir, Bialpirina, Bialpirinia, Caprin, Cardioaspirin, Cardioaspirina, Cardiomagnyl, Claradin, Colfarit, Colsprin, Contrheuma Retard, Coraspin, Coricidin, Coricidin D, Crystar, Darvon Compound, Dolean pH 8, Dominal, Duramax, Durlaza, ECM, Easprin, Ecopirin, Ecosprin, Ecotrin, Empirin, Endosprin, Endydol, Entericin, Enterophen, Enterosarine, Entrophen, Ewin, Extren, Gelprin, Globentyl, Globoid, Helicon, IWN 11, Idragin, Istopirin, Kapsazal, Lysoprin (pharmaceutical), Magnecyl, Measurin, Medisyl, Melhoral, Micristin, Miniasal, Mycropyrin, NSC 27223, NSC 406186, Neuronika, Novid, Nu-seals, O-Acetylsalicylic acid, Persistin, Polopiryna, Rheumintabletten, Rhodine 2312, Rhodine 3118, Rhodine 3220, Rhodine NC RP, Rhonal, Rivo, Rodine 3220, SP 189, Salacetin, Salcetogen, Saletin, Salospir, Salycylacetylsalicylic acid, Solpyron, Supac, Temperal, ThromboASS, Toldex, Triple-sal, Trombyl, Xaxa, Yasta, Zorprin, Zosprin, o-(Acetyloxy)benzoic acid, o-Acetoxybenzoic acid, o-Carboxyphenyl acetate
InChI Formula InChI=1S/C9H8O4/c1-6(10)13-8-5-3-2-4-7(8)9(11)12/h2-5H,1H3,(H,11,12)
SMILES CC(=O)Oc1ccccc1C(=O)O
IUPAC Name 2-acetyloxybenzoic acid
Molecular Weight (g/mol) 180.16
Formula Weight 180.0423
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.