missing translation for 'onlineSavingsMsg'
Learn More

Acetylsalicylic Acid, Mikromol™

Product Code. 17964066
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17964066 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17964066 Supplier Mikromol Supplier No. MM0133.000250
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Acetylsalicylic Acid
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Acetylsalicylic Acid
CAS 50-78-2
Analyte or Component Names Acetylsalicylic Acid
Shipping Condition Room Temperature
Molecular Formula C9 H8 O4
Certifications/Compliance ISO 17034
Synonym 2-Acetoxybenzoic Acid, 2-(Acetyloxy)benzoic Acid, Acylpyrin, Angettes, Aspro, Aspirin, ASA, Acetylsalicylic Acid, Benzoic acid, 2-(acetyloxy)-, Rhodine (7CI), Salicylic acid acetate (8CI), 2-(Acetyloxy)benzoic acid, 2-Acetoxybenzoic acid, 2-Carboxyphenyl acetate, A.S.A. Empirin, AC 5230, Acenterine, Acesal, Acesan, Acetard, Aceticyl, Acetilum acidulatum, Acetisal, Acetol, Acetonyl, Acetophen, Acetosal, Acetosalic acid, Acetosalin, Acetylin, Acetylsal, Acetylsalicylic acid, Acetyonyl, Acetysal, Acidum acetylsalicylicum, Acimetten, Acisal, Adiro, Albyl E, Asaflow, Asagran, Asatard, Ascoden 30, Ascolong, Ascriptin, Aspalon, Aspergum, Aspirdrops, Aspirin Protect 100, Aspirin Protect 300, Aspirin-Direkt, Aspirina 03, Aspro Clear, Aspropharm, Asteric, Astrix, Bayer, Benaspir, Bialpirina, Bialpirinia, Caprin, Cardioaspirina, Claradin, Colfarit, Colsprin, Contrheuma Retard, Coricidin, Coricidin D, Crystar, Darvon Compound, Dolean pH 8, Dominal, Doril, Duramax, ECM, Easprin, Ecosprin, Ecotrin, Empirin, Endosprin, Endydol, Entericin, Enterophen, Enterosarine, Entrophen, Ewin, Extren, Gelprin, Globentyl, Globoid, Helicon, Idragin, Istopirin, Kapsazal, Lysoprin (pharmaceutical), Magnecyl, Measurin, Medisyl, Melhoral, Micristin, Miniasal, Mycropyrin, NSC 27223, NSC 406186, Neuronika, Novid, Nu-seals, O-Acetylsalicylic acid, Persistin, Polopiryna, Rheumintabletten, Rhodine 2312, Rhodine NC RP, Rhonal, SP 189, Salacetin, Salcetogen, Saletin, Salospir, Salycylacetylsalicylic acid, Solpyron, Supac, Temperal, Toldex, Triple-sal, Trombyl, Xaxa, Yasta, Zorprin, o-(Acetyloxy)benzoic acid, o-Acetoxybenzoic acid, o-Carboxyphenyl acetate
InChI Formula InChI=1S/C9H8O4/c1-6(10)13-8-5-3-2-4-7(8)9(11)12/h2-5H,1H3,(H,11,12)
SMILES CC(=O)Oc1ccccc1C(=O)O
IUPAC Name 2-acetyloxybenzoic acid
Molecular Weight (g/mol) 180.16
Quantity 250 mg
Formula Weight 180.0423 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.