missing translation for 'onlineSavingsMsg'
Learn More

Acetylsalicylic Acid, LoGiCal

Product Code. 30457991
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30457991 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30457991 Supplier LoGiCal Supplier No. LGCFOR0133.00
 Restricted Item
Estimated Shipment: 29-09-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Acetylsalicylic Acid
CAS 50-78-2
Analyte or Component Names Acetylsalicylic Acid
Shipping Condition Room Temperature
Molecular Formula C9 H8 O4
Certifications/Compliance ISO 9001
Synonym 2-Acetoxybenzoic Acid, 2-(Acetyloxy)benzoic Acid, Acylpyrin, Angettes, Aspro, Aspirin, ASA, Acetylsalicylic Acid, Benzoic acid, 2-(acetyloxy)-, Rhodine (7CI), Salicylic acid acetate (8CI), 2-(Acetyloxy)benzoic acid, 2-Acetoxybenzoic acid, 2-Carboxyphenyl acetate, A.S.A. Empirin, AC 5230, Acenterine, Acesal, Acesan, Acetard, Aceticyl, Acetilum acidulatum, Acetisal, Acetol, Acetonyl, Acetophen, Acetosal, Acetosalic acid, Acetosalin, Acetylin, Acetylsal, Acetylsalicylic acid, Acetyonyl, Acetysal, Acidum acetylsalicylicum, Acimetten, Acisal, Adiro, Albyl E, Asaflow, Asagran, Asatard, Ascoden 30, Ascolong, Ascriptin, Aspalon, Aspergum, Aspirdrops, Aspirin Protect 100, Aspirin Protect 300, Aspirin-Direkt, Aspirina 03, Aspro Clear, Aspropharm, Asteric, Astrix, Bayer, Benaspir, Bialpirina, Bialpirinia, Caprin, Cardioaspirina, Claradin, Colfarit, Colsprin, Contrheuma Retard, Coricidin, Coricidin D, Crystar, Darvon Compound, Dolean pH 8, Dominal, Doril, Duramax, ECM, Easprin, Ecosprin, Ecotrin, Empirin, Endosprin, Endydol, Entericin, Enterophen, Enterosarine, Entrophen, Ewin, Extren, Gelprin, Globentyl, Globoid, Helicon, Idragin, Istopirin, Kapsazal, Lysoprin (pharmaceutical), Magnecyl, Measurin, Medisyl, Melhoral, Micristin, Miniasal, Mycropyrin, NSC 27223, NSC 406186, Neuronika, Novid, Nu-seals, O-Acetylsalicylic acid, Persistin, Polopiryna, Rheumintabletten, Rhodine 2312, Rhodine NC RP, Rhonal, SP 189, Salacetin, Salcetogen, Saletin, Salospir, Salycylacetylsalicylic acid, Solpyron, Supac, Temperal, Toldex, Triple-sal, Trombyl, Xaxa, Yasta, Zorprin, o-(Acetyloxy)benzoic acid, o-Acetoxybenzoic acid, o-Carboxyphenyl acetate
InChI Formula InChI=1S/C9H8O4/c1-6(10)13-8-5-3-2-4-7(8)9(11)12/h2-5H,1H3,(H,11,12)
Solution Type Neat
SMILES CC(=O)Oc1ccccc1C(=O)O
IUPAC Name 2-acetyloxybenzoic acid
Industry Type Clinical Testing
Molecular Weight (g/mol) 180.16
Quantity 10 mg
Formula Weight 180.0423 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.