missing translation for 'onlineSavingsMsg'
Learn More

Acetylpyrazine, MedChemExpress

Product Code. 16449326
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16449326 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16449326 Supplier Medchem Express Supplier No. HYW007692

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Acetylpyrazine (2-Acetylpyrazine) is used to form many polycyclic compounds, as useful structures in pharmaceuticals and perfumes. Acetylpyrazine is a component of the folates (vitamin B compounds).

Acetylpyrazine (2-Acetylpyrazine) is used to form many polycyclic compounds, as useful structures in pharmaceuticals and perfumes. Acetylpyrazine is a component of the folates (vitamin B compounds).

Specifications

Chemical Name or Material Acetylpyrazine
CAS 22047-25-2
Color White to Off-White
Purity Grade Notes Research
Molecular Formula C6H6N2O
Quantity 100 mg
Synonym 2-Acetylpyrazine
Solubility Information DMSO : 100 mg/mL (818.80 mM; Need ultrasonic)
SMILES C1=C(N=CC=N1)C(=O)C
Molecular Weight (g/mol) 122.13
Formula Weight 122.13
Percent Purity 99.95%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.