missing translation for 'onlineSavingsMsg'
Learn More

(Acetylmethylene)triphenylphosphorane, 99%

Product Code. 11450243
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11450243 10 g 10g
11460243 50 g 50g
11470243 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11450243 Supplier Thermo Scientific Alfa Aesar Supplier No. A11410.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1439-36-7 | C21H19OP | 318.36 g/mol

(Acetylmethylene)triphenylphosphorane is used as a Wittig reagent in synthetic chemistry, especially for the synthesis of functionalized pyrrolidines and cyclobutanones. It plays as a vital role in asymmetric allylboration for enantioselective synthesis of (+)-awajanomycin.It is also employed as a reactant in the preparation of 1,2-dioxanes with antitrypanosomal activity. Further, it is used in the preparation of amphibian pyrrolizidine alkaloids through allylic aminations and silicon-containing acyclic dienone musk odorants. In addition to this, it is involved in Domino Suzuki/Heck coupling reactions to prepare fluorenylidenes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(Acetylmethylene)triphenylphosphorane is used as a Wittig reagent in synthetic chemistry, especially for the synthesis of functionalized pyrrolidines and cyclobutanones. It plays as a vital role in asymmetric allylboration for enantioselective synthesis of (+)-awajanomycin.It is also employed as a reactant in the preparation of 1,2-dioxanes with antitrypanosomal activity. Further, it is used in the preparation of amphibian pyrrolizidine alkaloids through allylic aminations and silicon-containing acyclic dienone musk odorants. In addition to this, it is involved in Domino Suzuki/Heck coupling reactions to prepare fluorenylidenes.

Solubility
Soluble in chloroform. Slightly soluble in methanol.

Notes
Air sensitive. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1439-36-7
Molecular Formula C21H19OP
Molecular Weight (g/mol) 318.36
MDL Number MFCD00008774
InChI Key KAANTNXREIRLCT-UHFFFAOYSA-N
Synonym acetylmethylene triphenylphosphorane, 1-triphenylphosphoranylidene propan-2-one, 1-triphenylphosphoranylidene-2-propanone, 1-triphenylphosphoranylidene acetone, 2-propanone, 1-triphenylphosphoranylidene, acetonylidenetriphenyl phosphorane, acetylmethylene triphenylphosphine, triphenylacetylmethylene phosphorane, 2-propanone, triphenylphosphoranylidene, triphenylphosphoranylidene-2-propanone
PubChem CID 15038
IUPAC Name 1-(triphenyl-$l^{5}-phosphanylidene)propan-2-one
SMILES CC(=O)C=P(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 204°C to 209°C
Quantity 10 g
Beilstein 750077
Sensitivity Air sensitive
Solubility Information Soluble in chloroform. Slightly soluble in methanol.
Formula Weight 318.36
Percent Purity 99%
Chemical Name or Material (Acetylmethylene)triphenylphosphorane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.