missing translation for 'onlineSavingsMsg'
Learn More

Acetylleucine, MedChemExpress

Product Code. 16431118
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16431118 5 g 5g
16422349 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16431118 Supplier Medchem Express Supplier No. HYB1442
 Restricted Item
Estimated Shipment: 18-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Acetylleucine (N-Acetyl-DL-leucine), orally active compound, can be used for the research of acute vestibular vertigo, cerebellar ataxia and nystagmus.

Acetylleucine (N-Acetyl-DL-leucine), orally active compound, can be used for the research of acute vestibular vertigo, cerebellar ataxia and nystagmus.

Specifications

Chemical Name or Material Acetylleucine
CAS 99-15-0
Color White
Purity Grade Notes Research
Molecular Formula C8H15NO3
Synonym N-Acetyl-DL-leucine
Solubility Information DMSO : 120 mg/mL (692.80 mM; Need ultrasonic) ∣H2O : 6.67 mg/mL (38.51 mM; Need ultrasonic)
SMILES CC(C)CC(C(O)=O)NC(C)=O
Molecular Weight (g/mol) 173.21
Quantity 5 g
Formula Weight 173.21
Percent Purity 98.0%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.