missing translation for 'onlineSavingsMsg'
Learn More

Acetylenedicarboxylic acid, 97%

Product Code. 11423018
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11423018 25 g 25g
11413018 5 g 5g
11433018 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11423018 Supplier Thermo Scientific Alfa Aesar Supplier No. A18736.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 142-45-0 | C4H2O4 | 114.06 g/mol

Acetylenedicarboxylic acid is employed in the preparation of dimethyl acetylenedicarboxylate and potassium hydrogenacetylenedicarboxylate. It is also used as a laboratory reagent. Further, it is used as a precursor in organic synthesis and also serves as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Acetylenedicarboxylic acid is employed in the preparation of dimethyl acetylenedicarboxylate and potassium hydrogenacetylenedicarboxylate. It is also used as a laboratory reagent. Further, it is used as a precursor in organic synthesis and also serves as a pharmaceutical intermediate.

Solubility
Soluble in diethyl ether and water.

Notes
Air sensitive. Store in a cool place. Incompatible with bases, alkali, reducing agents and oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 142-45-0
Molecular Formula C4H2O4
Molecular Weight (g/mol) 114.06
MDL Number MFCD00004362
InChI Key YTIVTFGABIZHHX-UHFFFAOYSA-N
Synonym acetylenedicarboxylic acid, 2-butynedioic acid, butynedioic acid, acetylenedicarboxylate, unii-2d2oj4ko44, acetylenedicarboxylic acid 8ci, acetylendicarboxylate, hoocc=ccooh, acetylenedicarboxylicacid, pubchem13735
PubChem CID 371
ChEBI CHEBI:30781
IUPAC Name but-2-ynedioic acid
SMILES OC(=O)C#CC(O)=O

Specifications

Melting Point ∼182°C (decomposition)
Flash Point 131°C (267°F)
Quantity 25 g
UN Number UN2928
Beilstein 878357
Solubility Information Soluble in diethyl ether and water.
Formula Weight 114.06
Percent Purity 97%
Chemical Name or Material Acetylenedicarboxylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.