missing translation for 'onlineSavingsMsg'
Learn More

Acetylene-PEG4-maleimide

Product Code. 15486609
Click to view available options
Quantity:
25 mg
100 mg
500 mg
Unit Size:
100mg
25mg
500mg
This item is not returnable. View return policy

Product Code. 15486609

Brand: Thermo Scientific Alfa Aesar J64859.MB

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1609651-90-2 | C18H26N2O7 | 382.41 g/mol

Heterobifunctional cross linker enabling the connection of an azide containing compound or biomolecule to one with a thiol. The maleimide group will react preferentially with thiols to form a covalent adduct and the terminal alkyne will react with azides via copper-catalyzed 1,3-dipolar cycloaddition click chemistry. The hydrophilic PEG spacer adds to the water solubility of this reagent, allowing for easy modification cysteine containing biomolecules such as antibodies, proteins and peptides with the terminal alkyne. The PEG spacer also reduces aggregation of modified proteins stored in solution by adding hydrophilic character. A bifunctional linker with sulfhydryl reactive and azide reactive groups. Contains a PEG spacer to increase solubility

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Heterobifunctional cross linker enabling the connection of an azide containing compound or biomolecule to one with a thiol. The maleimide group will react preferentially with thiols to form a covalent adduct and the terminal alkyne will react with azides via copper-catalyzed 1,3-dipolar cycloaddition click chemistry. The hydrophilic PEG spacer adds to the water solubility of this reagent, allowing for easy modification cysteine containing biomolecules such as antibodies, proteins and peptides with the terminal alkyne. The PEG spacer also reduces aggregation of modified proteins stored in solution by adding hydrophilic character. A bifunctional linker with sulfhydryl reactive and azide reactive groups. Contains a PEG spacer to increase solubility

Solubility
Soluble in water.

Notes
Keep container tightly closed. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1609651-90-2
Molecular Formula C18H26N2O7
Molecular Weight (g/mol) 382.41
MDL Number MFCD22380740
InChI Key GHEBECZRHJXPTL-UHFFFAOYSA-N
Synonym acetylene-peg4-maleimide, propargyl-peg4-maleimide, alkyne-peg4-maleimide hplc
PubChem CID 91757777

Specifications

Color Colorless to Yellow
Quantity 25 mg
Solubility Information Soluble in water.
Formula Weight 382.41
Physical Form Viscous Liquid or Solid
Chemical Name or Material Acetylene-PEG4-maleimide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.