missing translation for 'onlineSavingsMsg'
Learn More

Acetylene-PEG4-biotin conjugate, Thermo Scientific™

Product Code. p-7043109
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15406979 25 mg 25mg
15354991 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15406979 Supplier Thermo Scientific Alfa Aesar Supplier No. J65577.MB
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

An acetylene-biotin conjugate with PEG spacer

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1262681-31-1
Molecular Formula C21H35N3O6S
Molecular Weight (g/mol) 457.59
MDL Number MFCD22380755
InChI Key SKMJWNZZFUDLKQ-UHFFFAOYNA-N
Synonym biotin-peg4-alkyne, acetylene-peg4-biotin conjugate
PubChem CID 75409985

Specifications

Color White
Quantity 25 mg
Formula Weight 457.59
Physical Form Solid
Chemical Name or Material Acetylene-PEG4-biotin conjugate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.