missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Acetylcholinesterase Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA586086
This item is not returnable.
View return policy
Description
Purity is > 95% by SDS-PAGE.
Acetylcholinesterase hydrolyzes the neurotransmitter, acetylcholine at neuromuscular junctions and brain cholinergic synapses, and thus terminates signal transmission. The Protein is also found on the red blood cell membranes, where it constitutes the Yt blood group antigen. Acetylcholinesterase exists in multiple molecular forms which possess similar catalytic properties, but differ in their oligomeric assembly and mode of cell attachment to the cell surface. The major form of acetylcholinesterase found in brain, muscle and other tissues is the hydrophilic species, which forms disulfide-linked oligomers with collagenous, or lipid-containing structural subunits.
Specifications
| Acetylcholinesterase | |
| Polyclonal | |
| Unconjugated | |
| ACHE | |
| ACE-1; ACEE; Acetylcholinesterase; acetylcholinesterase (Cartwright blood group); acetylcholinesterase (Yt blood group); acetylcholinesterase T-subunit; ACHE; ache protein; apoptosis-related acetylcholinesterase; ARACHE; EC 3.1.1.7; glycolipid-anchored form of acetylcholinesterase; N-ACHE; OTTHUMP00000211349; OTTHUMP00000211356; YT; Yt blood group; zgc:92550; ziehharmonika; zim | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 11423, 43, 83817 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| P21836, P22303, P37136 | |
| ACHE | |
| A synthetic peptide corresponding to residues in Human AChE. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction