missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetylcholinesterase/ACHE Antibody [Janelia Fluor™ 635], Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NB600-911JF635
This item is not returnable.
View return policy
Description
Acetylcholinesterase/ACHE Polyclonal antibody specifically detects Acetylcholinesterase/ACHE in Bovine samples. It is validated for ELISA,Immunoprecipitation,Western Blot
Specifications
| Acetylcholinesterase/ACHE | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Goat | |
| Multi-step | |
| RUO | |
| Primary | |
| Bovine | |
| Purified |
| ELISA, Immunoprecipitation, Western Blot | |
| Janelia Fluor 635 | |
| acetylcholinesterase, acetylcholinesterase (Yt blood group), AChE, apoptosis-related acetylcholinesterase, ARACHE, EC 3.1.1, EC 3.1.1.7, N-ACHE, YT, Yt blood group | |
| Acetylcholinesterase/ACHE [Bovine Erythrocytes] (Uniprot: P23795) | |
| 0.1 mL | |
| Cellular Markers, Neuronal Cell Markers, Neuroscience, Neurotransmission | |
| 43 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction