missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetylcholinesterase/ACHE Antibody, Alexa Fluor™ 350, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NB600-911AF350
This item is not returnable.
View return policy
Description
Acetylcholinesterase/ACHE Polyclonal specifically detects Acetylcholinesterase/ACHE in Bovine samples. It is validated for Western Blot, ELISA.
Specifications
| Acetylcholinesterase/ACHE | |
| Polyclonal | |
| Western Blot, ELISA | |
| acetylcholinesterase, acetylcholinesterase (Yt blood group), AChE, apoptosis-related acetylcholinesterase, ARACHE, EC 3.1.1, EC 3.1.1.7, N-ACHE, YT, Yt blood group | |
| Acetylcholinesterase/ACHE [Bovine Erythrocytes] (Uniprot: P23795) | |
| 0.1 mL | |
| Cellular Markers, Neuronal Cell Markers, Neuroscience, Neurotransmission | |
| 43 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, ELISA | |
| Alexa Fluor 350 | |
| 50mM Sodium Borate | |
| Goat | |
| Multi-step | |
| RUO | |
| Primary | |
| Bovine | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction