missing translation for 'onlineSavingsMsg'
Learn More

Acetylcholine iodide, 99%

Product Code. p-49839 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10649974 10 g 10g
10576933 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10649974 Supplier Thermo Scientific Acros Supplier No. 400180100
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2260-50-6 | C7H16INO2 | 273.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2260-50-6
Molecular Formula C7H16INO2
Molecular Weight (g/mol) 273.11
MDL Number MFCD00011815
InChI Key SMBBQHHYSLHDHF-UHFFFAOYSA-M
Synonym acetylcholine iodide, acetylcolina, 2-acetoxy-n,n,n-trimethylethanaminium iodide, acetylcolina italian, unii-7zcp12s7hq, choline, iodide, acetate ester, 2-acetoxyethyl trimethylammonium iodide, 7zcp12s7hq, ethanaminium, 2-acetyloxy-n,n,n-trimethyl-, iodide, 2-acetyloxy-n,n,n-trimethylethanamium iodide
PubChem CID 75271
SMILES [I-].CC(=O)OCC[N+](C)(C)C

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 162.0°C to 164.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (Argentometry)
Packaging Glass bottle
Linear Formula CH3COOCH2CH2N(CH3)3I
Quantity 10 g
Solubility Information Solubility in water: negligible.
IUPAC Name 2-acetyloxyethyl(trimethyl)azanium;iodide
Formula Weight 273.1
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material Acetylcholine iodide, 99%
Show More Show Less
compliance-icons
Product Identifier
  • Acetylcholine iodide
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.