missing translation for 'onlineSavingsMsg'
Learn More
Learn More
acetylated Tubulin(Lys40) Mouse anti-Canine, Human, Mouse, Porcine, Rat, Clone: 7E5H8, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66200-1-IG-20UL
This item is not returnable.
View return policy
Description
This antibody is specific to the acetylated tubulin; it does not recognize non-acetylated tubulin.
Alpha Tubulin is a part of a heterodimer consisting of alpha and beta tubulin subunits. The alpha and beta tubulins, which are each about 55 kDa MW, are homologous but not identical. Alpha-beta tubulin heterodimer is the basic building block of microtubules, and this intracellular cylindrical filamentous structure is present in almost eukaryotic cells. Microtubules serve as structural supports and lines of transport within the cell, as well as serving a key role in mitosis. Microtubules of the eukaryotic cytoskeleton perform essential and diverse functions and are composed of a heterodimer of alpha and beta tubulins. The alpha and beta tubulins represent the major components of microtubules, while gamma tubulin plays a critical role in the nucleation of microtubule assembly. There are multiple alpha and beta tubulin genes, which are highly conserved among species. The gene that encodes alpha tubulin is highly similar to the mouse and rat Tuba1 gene. Northern blotting studies have shown that the gene expression is predomitly found in morphologically differentiated neurologic cells.Specifications
| acetylated Tubulin(Lys40) | |
| Monoclonal | |
| 0.7 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P02550, P68369, P68370, Q71U36 | |
| LOC106557476, Tuba1a | |
| Peptide | |
| 20 μL | |
| Primary | |
| Pig, Rat, Canine, Human, Mouse | |
| Antibody | |
| IgG1 |
| Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 7E5H8 | |
| Unconjugated | |
| TUBA1A | |
| acetylated alpha tubulin, Acetylated tubulin, Acetylated tubulin (Lys40), acetylated-a-tubulin, acetyl-a-tubulin, alpha tubulin, Alpha tubulin 3, tubulin | |
| Mouse | |
| Protein G | |
| RUO | |
| 100135051, 106557476, 22142, 64158, 7846 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction