missing translation for 'onlineSavingsMsg'
Learn More

Acetylacetonatobis(ethylene)rhodium(I), TRC

Product Code. 30315606
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30315606 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30315606 Supplier Toronto Research Chemicals Supplier No. TRCA1711935MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Acetylacetonatobis(ethylene)rhodium(I)
CAS 12082-47-2
Molecular Formula C9 H11 O2 Rh
Quantity 5 mg
Synonym 2,4-Pentanedionatobis(ethylene)rhodium, 2,4-Pentanedionatobis(ethylene)rhodium(I), Acetylacetonatobis(ethylene)rhodium(I), Bis(ethylene)(2,4-pentanedionato)rhodium, Bis(ethylene)rhodium(I) Acetylacetonate, Bisethylenerhodium Acetylacetonate, Diethenylrhodium Acetylacetonate, Diethylene(acetylacetonato)rhodium, bis(eta2-ethene)(2,4-pentanedionato-kO,kO')rhodiumn
InChI Formula InChI=1S/C5H7O2.2C2H2.Rh/c1-4(6)3-5(2)7;2*1-2;/h3H,1-2H3;2*1-2H;/q-1;;;+1
SMILES CC1=[O][Rh+]23(C=C2)(C=C3)[O]=C(C)[CH-]1
Molecular Weight (g/mol) 254.088
Formula Weight 253.981
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.