missing translation for 'onlineSavingsMsg'
Learn More

(Acetylacetonato)dicarbonylrhodium(I), TRC

Product Code. 30322994
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30322994 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30322994 Supplier Toronto Research Chemicals Supplier No. TRCA168710500MG
 Restricted Item
Estimated Shipment: 05-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (Acetylacetonato)dicarbonylrhodium(I)
CAS 14874-82-9
Molecular Formula C7H7O4Rh
Quantity 500 mg
Synonym Acetylacetonatorhodium Dicarbonyl, Dicarbonyl(2,4-pentanedionato)rhodium, Dicarbonyl(2,4-pentanedionato-κO,κO')rhodium, Dicarbonylacetylacetonatorhodium, Dicarbonylacetylacetonatorhodium(I), Dicarbonylrhodium Acetylacetonate, Dicarbonylrhodium(I) Acetylacetonate, Rh(acac)(CO)2, Rhodium Acetylacetonate dicarbonyl, Rhodium Biscarbonyl Acetylacetonate, Rhodium Dicarbonylacetylacetonate, Rhodium (2,4-Pentanedionato)dicarbonyl, Rhodium Dicarbonyl(2,4-pentanedionato), Rhodium Dicarbonyl(2,4-pentanedionato-O,O')-(SP-4-2), Rhodium Dicarbonyl(2,4-pentanedionato-κO,κO')-(SP-4-2), (SP-4-2)-Dicarbonyl(2,4-pentanedionato-κO2,κO4)rhodium, (Acetylacetonate)dicarbonylrhodium, (Acetylacetonato)dicarbonyl Rhodium Acetylacetonatobis(carbonyl)rhodium
SMILES CC(/C=C(O[Rh])/C)=O.C=O.C=O
Molecular Weight (g/mol) 258.03
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.