missing translation for 'onlineSavingsMsg'
Learn More

Acetyl-β-methylcholine bromide, 99%, Thermo Scientific Chemicals

Product Code. 10368170
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 333-31-3
Molecular Formula C8H18BrNO2
Molecular Weight (g/mol) 240.14
MDL Number MFCD00011816
InChI Key MMVPLEUBMWUYIB-UHFFFAOYNA-M
Synonym methacholine bromide, 2-acetoxy-n,n,n-trimethylpropan-1-aminium bromide, acetyl beta-methylcholine bromide, 1-propanaminium, 2-acetyloxy-n,n,n-trimethyl-, bromide, 1-propanaminium, 2-acetyloxy-n,n,n-trimethyl-, bromide 1:1, methacholini bromidum, methacholine bromide nf, c8h18no2.br, mecholyl bromide, methacholinebromide
PubChem CID 92754
IUPAC Name 2-acetyloxypropyl(trimethyl)azanium;bromide
SMILES [Br-].CC(C[N+](C)(C)C)OC(C)=O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 150°C to 152°C
Color White
Loss on Drying 1% max.
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (on dried substance) (Argentometry)
Packaging Glass bottle
Linear Formula CH3CO2CH(CH3)CH2N(CH3)3Br
Quantity 5 g
Beilstein 04, III, 758
Merck Index 10, 5793
Solubility Information Solubility in water: soluble
Formula Weight 240.14
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material Acetyl-β-methylcholine bromide, 99%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'