missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetyl-Lysine Antibody (7F8), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB110-96876H
This item is not returnable.
View return policy
Description
Acetyl-Lysine Monoclonal antibody specifically detects Acetyl-Lysine in Nonspecies specific samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin).
Specifications
| Acetyl-Lysine | |
| Monoclonal | |
| HRP | |
| PBS with No Preservative | |
| Mouse | |
| Protein G purified | |
| Primary | |
| Non-species specific | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 7F8 | |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| acetyl Lysine, AC-LYS-OH, Lysine, N-acetyl lysine | |
| Acetylated KLH | |
| 0.1 mL | |
| Detects proteins containing acetylated lysine residues in ELISA and western blots. Does not detect non-acetylated lysine residues. | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction