missing translation for 'onlineSavingsMsg'
Learn More

Acetyl-L-glutamine, TRC

Product Code. 30327693
Change view
Click to view available options
Quantity:
500 mg
1 g
5 g
Unit Size:
1g
500mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30327693 1 g 1g
30322800 500 mg 500mg
30311093 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30327693 Supplier Toronto Research Chemicals Supplier No. TRCA2978601G
 Restricted Item
Estimated Shipment: 04-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-Acetyl-L-glutamine
CAS 2490-97-3
Molecular Formula C7H12N2O4
Synonym L-Glutamine, N2-acetyl- (9CI, ACI), Glutamine, N2-acetyl-, L- (6CI, 7CI, 8CI), N2-Acetyl-L-glutamine (ACI), (2S)-2-Acetamido-5-amino-5-oxopentanoic acid, (2S)-4-Carbamoyl-2-acetamidobutanoic acid, Aceglutamid, Aceglutamide, Acetylglutamine, Acutil S, N2-Acetylglutamine, NSC 186896, α-N-Acetyl-L-glutamine
InChI Formula InChI=1S/C7H12N2O4/c1-4(10)9-5(7(12)13)2-3-6(8)11/h5H,2-3H2,1H3,(H2,8,11)(H,9,10)(H,12,13)/t5-/m0/s1
SMILES CC(=O)N[C@@H](CCC(=O)N)C(=O)O
IUPAC Name (2S)-2-acetamido-5-amino-5-oxo-pentanoic acid
Molecular Weight (g/mol) 188.18
Quantity 1 g
Formula Weight 188.0797
Shipping Condition Room Temperature
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.