Learn More
Invitrogen™ H2BK23ac Recombinant Rabbit Monoclonal Antibody (RM260)
Rabbit Recombinant Monoclonal Antibody
Marque: Invitrogen™ MA527885
Description
Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.
Histones are basic nuclear proteins that are responsible for the nucleosome structure of the chromosomal fiber in eukaryotes. Nucleosomes consist of approximately 146 bp of DNA wrapped around a histone octamer composed of pairs of each of the four core histones (H2A, H2B, H3, and H4). The nucleosomes wrap further and compact DNA into chromotin, limiting DNA accessibility to the cellular machineries which require DNA as a template. Histones play a central role in transciption regulation, DNA repair, DNA replication, and chromosomal stability. H2B has a broad antibacterial activity.
Spécification
| H2BK23ac | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.09% sodium azide | |
| A0A2R8Y619, O60814, P06899, P0C1H6, P23527, P33778, P57053, P58876, P62807, Q16778, Q5QNW6, Q7Z2G1, Q8N257, Q93079, Q96A08, Q99877, Q99879, Q99880 | |
| H2B, H2bc1, H2BC10, H2BC11, H2BC12, H2BC13, H2BC14, H2BC15, H2BC17, H2bc18, H2BC21, H2BC3, H2BC4, H2BC5, H2BC8, H2BC9, H2BE1, H2BU1, H2BW1, H2BW2, LOC102724334 | |
| An acetyl-peptide corresponding to Acetyl-Histone H2B (Lys23). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| ELISA, Western Blot, Luminex, Immunocytochemistry | |
| RM260 | |
| Unconjugated | |
| H2BW2 | |
| Acetyl-Histone H2B K23; Acetyl-Histone H2B Lys23; Acetyl-Histone H3 K23; Acetyl-Histone H3 Lys23; bA317E16.3; dJ160A22.3; dJ193B12.2; dJ221C16.3; dJ221C16.6; dJ221C16.8; dJ97D16.4; GL105; H2B; H2B histone family member M; H2B histone family member W testis-specific; H2B histone family member W, testis specific; H2B histone family, member A; H2B histone family, member B; H2B histone family, member C; H2B histone family, member D; H2B histone family, member E; H2B histone family, member F; H2B histone family, member J; H2B histone family, member K; H2B histone family, member L; H2B histone family, member M; H2B histone family, member N; H2B histone family, member Q; H2B histone family, member R; H2B histone family, member T; H2B histone family, member U, (testis-specific); H2B histone family, member W, testis-specific; H2B K; H2B type 12; H2B.1; H2B.1A; H2B.1B; H2B.2; H2B/a; H2B/b; H2B/c; H2B/d; H2B/e; H2B/f; H2B/g; H2B/h; H2B/j; H2B/k; H2B/l; H2B/n; H2B/q; H2B/r; H2B/S; H2Bb; H2BC10; H2BC4; H2BC6; H2BC7; H2BC8; H2BFA; H2BFAiii; H2BFB; H2BFC; H2BFD; H2BFE; H2BFF; H2BFG; H2BFH; H2BFJ; H2BFK; H2BFL; H2BFM; H2BFN; H2BFQ; H2BFR; H2BFS; H2BFT; H2BFU; H2BFWT; H2BGL105; H2BJ; H2BK; H2BLys23ac; H2BQ; HIRA-interacting protein 1; HIRA-interacting protein 2; HIRIP1; HIRIP2; HIST1H2BA; HIST1H2BB; HIST1H2BC; HIST1H2BD; HIST1H2BE; HIST1H2BF; HIST1H2BG; HIST1H2BH; HIST1H2BI; HIST1H2BJ; HIST1H2BK; HIST1H2BL; HIST1H2BM; HIST1H2BN; HIST1H2BO; HIST2H2BE; HIST2H2BF; HIST3H2BB; histone 1, H2ba; histone 1, H2bb; histone 1, H2bc; histone 1, H2bd; histone 1, H2bg; histone 1, H2bh; histone 1, H2bi; histone 1, H2bj; histone 1, H2bk; histone 1, H2bl; histone 1, H2bm; histone 1, H2bn; histone 1, H2bo; histone 2, H2be; histone 3, H2bb; histone cluster 1, H2ba; histone cluster 1, H2bb; histone cluster 1, H2bc; histone cluster 1, H2bd; histone cluster 1, H2bg; histone cluster 1, H2bh; histone cluster 1, H2bi; histone cluster 1, H2bj; histone cluster 1, H2bk; histone cluster 1, H2bl; histone cluster 1, H2bm; histone cluster 1, H2bn; histone cluster 1, H2bo; histone cluster 2, H2be; histone cluster 2, H2bf; histone cluster 3, H2bb; histone family member; histone H2B type 1-A; Histone H2B type 1-B; histone H2B type 1-C/E/F/G/I; Histone H2B type 1-D; histone H2B type 1-H; Histone H2B type 1-J; histone H2B type 1-K; Histone H2B type 1-L; histone H2B type 1-M; Histone H2B type 1-N; histone H2B type 1-O; Histone H2B type 2-E; histone H2B type 2-F; Histone H2B type 3-B; histone H2B type F-M; Histone H2B type F-S; histone H2B type F-S-like; histone H2B type W-T; Histone H2B, testis; Histone H2B.1; histone H2B.1 A; Histone H2B.1 B; histone H2B.2; Histone H2B.a; histone H2B.b; Histone H2B.c; histone H2B.d; Histone H2B.e; histone H2B.f; Histone H2B.g; Histone H2B.h; Histone H2B.j; histone H2B.k; Histone H2B.l; histone H2B.n; Histone H2B.q; histone H2B.r; Histone H2B.s; histone H2B-GL105; LOC102724334; STBP; Testis-specific histone H2B; TH2B; TSH2B; TSH2B.1 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 102724334, 114483833, 128312, 158983, 255626, 286436, 3017, 3018, 440689, 8339, 8340, 8341, 8342, 8345, 8346, 8347, 8348, 8349, 85236, 8970 | |
| -20°C | |
| Liquid |
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.