missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetyl Coenzyme A Carboxylase Beta Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
£292.00 - £410.00
Specifications
| Antigen | Acetyl Coenzyme A Carboxylase Beta |
|---|---|
| Concentration | 0.1mg/mL |
| Dilution | Western Blot 0.04-0.4 ug/ml, Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18452242
|
Novus Biologicals
NBP1-90274-25ul |
25 μL |
£292.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18240856
|
Novus Biologicals
NBP1-90274 |
0.1 mL |
£410.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Acetyl Coenzyme A Carboxylase Beta Polyclonal specifically detects Acetyl Coenzyme A Carboxylase Beta in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| Acetyl Coenzyme A Carboxylase Beta | |
| Western Blot 0.04-0.4 ug/ml, Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| O00763 | |
| 32 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:ITKSKSEANLIPSQEPFPASDNSGETPQRNGEGHTLPKTPSQAEPASHKGPKDAGRRRNSLPPSHQKPPRNPLSSSDAAPSPELQANGTGTQGLEATDTNGLSSSARPQGQQAGSPSKEDKKQANIKRQLMTNFILGSFDDYSS | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 0.1mg/mL | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| ACC2ACC-beta, ACCBacetyl-CoA carboxylase 2, acetyl-CoA carboxylase beta, acetyl-Coenzyme A carboxylase beta, EC 6.4.1.2, HACC275 | |
| ACACB | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title