missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Acetyl-CoA Carboxylase Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA599547
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total ACC1.
Acetyl-CoA carboxylase (ACC) is a complex multifunctional enzyme system. ACC is a biotin-containing enzyme which catalyzes the carboxylation of acetyl-CoA to malonyl-CoA, the rate-limiting step in fatty acid synthesis. There are two ACC forms, alpha and beta, encoded by two different genes. ACC-alpha is highly enriched in lipogenic tissues. The enzyme is under long term control at the transcriptional and translational levels and under short term regulation by the phosphorylation/dephosphorylation of targeted serine residues and by allosteric transformation by citrate or palmitoyl-CoA. Multiple alternatively spliced transcript variants divergent in the 5' sequence and encoding distinct isoforms have been found for this gene.
Specifications
| Acetyl-CoA Carboxylase | |
| Polyclonal | |
| Unconjugated | |
| Acaca | |
| A1Z784_DROME; A530025K05Rik; Acac; Acaca; ACACAD; Acc; ACC1; ACCA; ACC-alpha; ACC-PA; ACC-PB; ACC-PC; ACC-PE; ACC-PF; ACC-PG; aceto-acetyl-CoA-thiolase; acetyl CoA carboxylase; acetyl coenzyme A carboxylase; Acetyl-CoA carboxylase; Acetyl-CoA carboxylase 1; acetyl-CoA carboxylase 265; acetyl-CoA carboxylase alpha; acetyl-CoA-carboxylase; acetyl-coenzyme A carboxylase; acetyl-coenzyme A carboxylase alpha; ACoT; Biotin carboxylase; CG11198; CG11198-PA; CG11198-PB; CG11198-PC; CG11198-PE; CG11198-PF; CG11198-PG; CG8723; dACC; DmACC; Dmel\CG11198; Dmel_CG11198; EC 6.3.4.14; EC 6.4.1.2; FBgn0043811; GH12002p; Gm738; I79_006999; unnamed protein product | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 104371, 107476, 31, 60581 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P11497, Q13085, Q5SWU9 | |
| Acaca, ACC | |
| A synthesized peptide derived from human ACACA(Accession Q13085), corresponding to amino acid residues L59-V109. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction