missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetyl-CoA Carboxylase alpha/ACACA, p Ser79 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-05407-20ul
This item is not returnable.
View return policy
Description
Acetyl-CoA Carboxylase alpha/ACACA Polyclonal antibody specifically detects Acetyl-CoA Carboxylase alpha/ACACA in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| Acetyl-CoA Carboxylase alpha/ACACA | |
| Polyclonal | |
| Western Blot 1:500 - 1:1000 | |
| ACACacetyl-CoA carboxylase 1, ACC1ACC, ACCA, ACC-alpha, acetyl-CoA carboxylase alpha, acetyl-Coenzyme A carboxylase alpha, EC 6.4.1.2 | |
| A synthetic phosphorylated peptide around S79 of human Acetyl-CoA Carboxylase alpha/ACACA (NP_942133.1). SMSGL | |
| 20 μg | |
| Breast Cancer, Lipid and Metabolism, Neuroscience, Phospho Specific | |
| 31 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction