missing translation for 'onlineSavingsMsg'
Learn More

Acetyl Chloride, TRC

Product Code. 30323367
missing translation for 'orderingAttributeHoverText'
Quantity:
100 g
missing translation for 'unitSize'
100g
This item is not returnable. View return policy

Product Code. 30323367

missing translation for 'mfr': Toronto Research Chemicals TRCA164496100G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Acetyl Chloride
CAS 75-36-5
Molecular Formula C2H3ClO
Quantity 100 g
Synonym Acetic Acid Chloride, Acetic Chloride, Ethanoyl Chloride, AcCl
InChI Formula InChI=1S/C2H3ClO/c1-2(3)4/h1H3
SMILES CC(=O)Cl
IUPAC Name acetyl chloride
Molecular Weight (g/mol) 78.5
Formula Weight 77.9872
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.