missing translation for 'onlineSavingsMsg'
Learn More

Acetyl Chloride, TRC

Product Code. 30328417
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30328417 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30328417 Supplier Toronto Research Chemicals Supplier No. TRCA16449610G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifikationer

Chemical Name or Material Acetyl Chloride
CAS 75-36-5
Molecular Formula C2H3ClO
Quantity 10 g
Synonym Acetic Acid Chloride, Acetic Chloride, Ethanoyl Chloride, AcCl
InChI Formula InChI=1S/C2H3ClO/c1-2(3)4/h1H3
SMILES CC(=O)Cl
IUPAC Name acetyl chloride
Molecular Weight (g/mol) 78.5
Formula Weight 77.9872
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.